Stampa…

Anti-SEMA6C Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fornitore: Bioss
BOSSBS-11483R-A555EA 858 EUR
BOSSBS-11483R-A555
Anti-SEMA6C Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorpi
Semaphorins are a family of cell surface and secreted proteins involved in neural development that are conserved from insects to humans. Members of this family are approximately 750 amino acids in length (including signal sequences) and are defined by a conserved extracellular “semaphorin” domain of approximately 500 amino acids containing 14-16 cysteines, blocks of conserved sequences and no obvious repeats. The transmembrane semaphorins are characterized by an additional 80 amino acid transmembrane domain and an 80-110 amino acid cytoplasmic domain. SEMA6C, also known as SEMA Y, is a transmembrane protein expressed in fetal brain and adult skeletal muscle. Three isoforms of this semaphorin exist due to alternative splicing: SEMA6C 1, SEMA6C 2 and SEMA6C 3. The extracellular domain of SEMA6C induces growth cone collapse of dorsal root ganglion and plays a role in generation or stability of entorhino-hippocampal synapses.

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SEMA6C
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR