Stampa…

Anti-ARF6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Fornitore: Bioss
BOSSBS-1145R-A350EA 858 EUR
BOSSBS-1145R-A350
Anti-ARF6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorpi
This gene encodes a member of the human ARF gene family, which is part of the RAS superfamily. The ARF genes encode small guanine nucleotide-binding proteins that stimulate the ADP-ribosyltransferase activity of cholera toxin and play a role in vesicular trafficking and as activators of phospholipase D. The product of this gene is localized to the plasma membrane, and regulates vesicular trafficking, remodelling of membrane lipids, and signaling pathways that lead to actin remodeling. A pseudogene of this gene is located on chromosome 7.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ARF6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR