Stampa…

Anti-SYNPR Rabbit Polyclonal Antibody (Cy3®)

Fornitore: Bioss
BOSSBS-11367R-CY3EA 866 EUR
BOSSBS-11367R-CY3
Anti-SYNPR Rabbit Polyclonal Antibody (Cy3®)
Anticorpi
Intrinsic membrane protein of small synaptic vesicles. Probable vesicular channel protein (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYNPR
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR