Stampa…

Anti-NOCT Rabbit Polyclonal Antibody (Cy5®)

Fornitore: Bioss
BOSSBS-11444R-CY5EA 858 EUR
BOSSBS-11444R-CY5
Anti-NOCT Rabbit Polyclonal Antibody (Cy5®)
Anticorpi
Nocturnin is a 431 amino acid circadian deadenylase protein that is expressed in a broad range of tissues with greatest abundance in the liver, kidney and testis. Nocturnin plays a role in circadian regulation as well as diet-induced obesity. The mRNA abundance of Nocturnin exhibits circadian rhythmicity, peaking after dusk in photoreceptors, spleen, heart, kidney and liver. Nocturnin is thought to be responisble for turning off genes that are involved in circadian regulation. In Xenopus retinal photoreceptor cells, the rhythmic regulation of Nocturnin is thought to be controlled by phosphorylated CREB. Mice lacking Nocturnin remain lean on high fat diets with a reduction in visceral fat, which suggests that this protein may also be responsible for lipid metabolism and fat storage.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOCT
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR