Stampa…

Anti-SLC16A2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Fornitore: Bioss
BOSSBS-11434R-A647EA 858 EUR
BOSSBS-11434R-A647
Anti-SLC16A2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorpi
Very active and specific thyroid hormone transporter. Stimulates cellular uptake of thyroxine (T4), triiodothyronine (T3), reverse triiodothyronine (rT3) and diidothyronine. Does not transport Leu, Phe, Trp or Tyr (By similarity).

Recommended Dilutions: Flow Cytometry: 1:20-100; IF(IHC-P): 1:50-200

Type: Primary
Antigen: SLC16A2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR