Stampa…

Anti-NG Rabbit Polyclonal Antibody (Cy5®)

Fornitore: Bioss
BOSSBS-11435R-CY5EA 858 EUR
BOSSBS-11435R-CY5
Anti-NG Rabbit Polyclonal Antibody (Cy5®)
Anticorpi
Acts as a "third messenger" substrate of protein kinase C-mediated molecular cascades during synaptic development and remodeling. Binds to calmodulin in the absence of calcium (By similarity).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NG/Neurogranin
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR