Stampa…

Anti-SLC16A2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Fornitore: Bioss
BOSSBS-11434R-PEEA 876 EUR
BOSSBS-11434R-PE
Anti-SLC16A2 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Anticorpi
Very active and specific thyroid hormone transporter. Stimulates cellular uptake of thyroxine (T4), triiodothyronine (T3), reverse triiodothyronine (rT3) and diidothyronine. Does not transport Leu, Phe, Trp or Tyr (By similarity).

Recommended Dilutions: Flow Cytometry: 1:20-100

Type: Primary
Antigen: SLC16A2
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR