Stampa…

Anti-SYNGR4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Fornitore: Bioss
BOSSBS-11375R-A350EA 858 EUR
BOSSBS-11375R-A350
Anti-SYNGR4 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorpi
The synaptogyrin family of proteins are integral membrane proteins containing four transmembrane regions. Synaptogyrins are tyrosine-phosphorylated proteins with two neuronal (Synaptogyrins 1 and 3) and one ubiquitous (Synaptogyrin-2) isoform. Synaptophysin and synaptogyrin represent the major constituents of synaptic vesicles. The 26kDa protein Synaptogyrin-1 is associated with presynaptic vesicles in neuronal cells. Synaptogyrin-2, also known as Cellugyrin has a tyrosine phosphorylated C-terminal cytoplasmic tail, and is involved in the regulation of membrane traffic in non-neuronal cells. Synaptogyrin-3 is expressed mainly in brain and placenta. The SYNGR4 gene encodes for the 234 amino acid protein Synaptogyrin-4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SYNGR4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR