Stampa…

Anti-RIMS1/ABCA4 Rabbit Polyclonal Antibody (Cy5®)

Fornitore: Bioss
BOSSBS-11356R-CY5EA 858 EUR
BOSSBS-11356R-CY5
Anti-RIMS1/ABCA4 Rabbit Polyclonal Antibody (Cy5®)
Anticorpi
The protein encoded by this gene is a RAS gene superfamily member that regulates synaptic vesicle exocytosis. This gene also plays a role in the regulation of voltage-gated calcium channels during neurotransmitter and insulin release. Mutations have suggested a role cognition and have been identified as the cause of cone-rod dystrophy type 7. Multiple transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq, Mar 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RIMS1/ABCA4
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR