Stampa…

Anti-CLSTN2 Rabbit Polyclonal Antibody (Cy3®)

Fornitore: Bioss
BOSSBS-11339R-CY3EA 858 EUR
BOSSBS-11339R-CY3
Anti-CLSTN2 Rabbit Polyclonal Antibody (Cy3®)
Anticorpi
Members of the calsyntenin protein family are localized to the post-synaptic membrane of exicitatory central nervous system (CNS) synapses. Calsyntenin-2, also known as Alcadein-gamma, is a 955 amino acid protein that localizes to the endoplasmic reticulum, golgi apparatus and plasma membranes. Containing 2 cadherin-like repeats in its N-terminal extracellular region, calsyntenin-2 binds synaptic calcium with its cytoplasmic domain, suggesting a role in the modulation of calcium-mediated postsynaptic signals. Under normal physiological conditions, calsyntenin-2 is protoeolytically processed in an event in which the primary zeta-cleavage generates a short C-terminal transmembrane fragment and a long extracellular N-terminal domain.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLSTN2
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR