Stampa…

Anti-MNX1 Rabbit Polyclonal Antibody (Cy5®)

Fornitore: Bioss
BOSSBS-11320R-CY5EA 866 EUR
BOSSBS-11320R-CY5
Anti-MNX1 Rabbit Polyclonal Antibody (Cy5®)
Anticorpi
Putative transcription factor involved in pancreas development and function.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MNX1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR