Stampa…

Anti-SAP102 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornitore: Bioss
BOSSBS-11315R-FITCEA 858 EUR
BOSSBS-11315R-FITC
Anti-SAP102 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
MPP3, also known as Discs large homolog 3 (DLG3), is a 585 amino acid protein belonging to the MAGUK family. Proteins in the MAGUK family, including CASK, ZO-1, ZO-2, p55 and MPP2, are known to regulate cell proliferation, intracellular junctions and signaling pathways by interacting with the cytoskeleton. MPP3 contains one guanylate kinase-like domain, one PDZ domain, two L27 domains and one SH3 domain. Via the PDZ domain, MMP3 interacts with the C-terminus of TSLC1. MMP3 has also been found to interact with SR-2A and SR-4.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SAP102
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR