Stampa…

Anti-INSL3 Rabbit Polyclonal Antibody (Cy3®)

Fornitore: Bioss
BOSSBS-11301R-CY3EA 858 EUR
BOSSBS-11301R-CY3
Anti-INSL3 Rabbit Polyclonal Antibody (Cy3®)
Anticorpi
This gene encodes a member of the insulin-like hormone superfamily. The encoded protein is mainly produced in gonadal tissues. Studies of the mouse counterpart suggest that this gene may be involved in the development of urogenital tract and female fertility. This protein may also act as a hormone to regulate growth and differentiation of gubernaculum, and thus mediating intra-abdominal testicular descent. Mutations in this gene may lead to cryptorchidism. Alternate splicing results in multiple transcript variants. [provided by RefSeq, May 2012].

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: INSL3
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR