Stampa…

Anti-5-HT Rabbit Polyclonal Antibody (Cy5.5®)

Fornitore: Bioss
BOSSBS-1126R-CY5.5EA 842 EUR
BOSSBS-1126R-CY5.5
Anti-5-HT Rabbit Polyclonal Antibody (Cy5.5®)
Anticorpi
Serotonin (pronounced) (5-hydroxytryptamine, or 5-HT) is a monoamine neurotransmitter synthesized in serotonergic neurons in the central nervous system (CNS) and enterochromaffin cells in the gastrointestinal tract of animals including humans. Serotonin is also found in many mushrooms and plants, including fruits and vegetables.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: 5-HT
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR