Stampa…

Anti-EPS15 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BOSSBS-11251R-CY7EA 858 EUR
BOSSBS-11251R-CY7
Anti-EPS15 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
Elucidation of the mechanism by which receptor tyrosine kinases (RTKs) modulate cellular physiology in response to stimuli is critical to the understanding of growth regulation. Miscues in RTK signaling pathways can result in cellular transformation and ultimately in cancer. Two novel EGF receptor substrates designated EGF-receptor pathway substrates 8 and 15, or Eps8 and Eps15, have been described. Eps8 and Eps15 are proteins, respectively that become tyrosine phosphorylated subsequent to EGF stimulation. Overexpression of Eps15 in NIH/3T3 cells causes cellular transformation, implying involvement in the regulation of cell proliferation. Eps15 is capable of binding the amino terminal portion of Crk via a conserved proline-rich domain, characteristic of all Crk binding proteins (5). Overexpression of Eps8 in both fibroblasts and hematopoietic cells results in an increased mitogenic response to EGF. Eps8 has been shown to associate with the EGF receptor despite its lack of a functional SH2 domain. Further characterization suggests the protein has both a PH domain and a SH3 domain, the functional significance of which are not yet known.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: EPS15
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR