Stampa…

Anti-JUNB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornitore: Bioss
BOSSBS-11228R-FITCEA 885 EUR
BOSSBS-11228R-FITC
Anti-JUNB Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Transcription factor involved in regulating gene activity following the primary growth factor response. Binds to the DNA sequence 5'-TGA[CG]TCA-3'.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: JUNB
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR