Stampa…

Anti-PRPH2 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BOSSBS-11197R-CY7EA 858 EUR
BOSSBS-11197R-CY7
Anti-PRPH2 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
May function as an adhesion molecule involved in stabilization and compaction of outer segment disks or in the maintenance of the curvature of the rim. It is essential for disk morphogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PRPH2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR