Stampa…

Anti-CEBPG Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Fornitore: Bioss
BOSSBS-11188R-A488EA 858 EUR
BOSSBS-11188R-A488
Anti-CEBPG Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorpi
Transcription factor that binds to the enhancer element PRE-I (positive regulatory element-I) of the IL-4 gene. Might change the DNA-binding specificity of other transcription factors and recruit them to unusual DNA sites.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CEBPG
Clonality: Polyclonal
Clone: 0
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR