Stampa…

Anti-MOBP Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fornitore: Bioss
BOSSBS-11184R-A555EA 858 EUR
BOSSBS-11184R-A555
Anti-MOBP Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorpi
May play a role in compacting or stabilizing the myelin sheath, possibly by binding the negatively charged acidic phospholipids of the cytoplasmic membrane.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MOBP
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR