Stampa…

Anti-PCDHGA9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornitore: Bioss
BOSSBS-11150R-FITCEA 858 EUR
BOSSBS-11150R-FITC
Anti-PCDHGA9 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Protocadherins are a large family of cadherin-like cell adhesion proteins that are involved in the establishment and maintenance of neuronal connections in the brain. There are three protocadherin (PCDH) gene clusters, designated alpha, beta and gamma, all of which contain multiple tandemly arranged genes. PCDHGA9 (Protocadherin gamma-A9) is a 932 amino acid that is one of 22 proteins encoded by the protocadherin gamma cluster. The protocadherein gamma cluster consists of three subfamilies (A, B and C) and PCDHGA9 is a member of the gamma subfamily A. PCDHGA9 is a type I transmembrane receptor containing six cadherin motifs and is expressed in the central nervous system where it localizes to synapses. Members of the gamma cluster of protocadherins are essential for neuronal survival. There are two isoforms of PCDHGA9 that are produced as a result of alternative splicing events.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCDHGA9
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR