Stampa…

Anti-NECTIN1 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BOSSBS-11126R-CY7EA 858 EUR
BOSSBS-11126R-CY7
Anti-NECTIN1 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
Promotes cell-cell contacts by forming homophilic or heterophilic trans-dimers. Heterophilic interactions have been detected between PVRL1/nectin-1 and PVRL3/nectin-3 and between PVRL1/nectin-1 and PVRL4/nectin-4. Functions as an entry receptor for herpes simplex virus and pseudorabies virus. Has some neurite outgrowth-promoting activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PVRL1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR