Stampa…

Anti-CNTNAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fornitore: Bioss
BOSSBS-11128R-A555EA 858 EUR
BOSSBS-11128R-A555
Anti-CNTNAP1 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorpi
Seems to play a role in the formation of functional distinct domains critical for saltatory conduction of nerve impulses in myelinated nerve fibers. Seems to demarcate the paranodal region of the axo-glial junction. In association with contactin may have a role in the signaling between axons and myelinating glial cells.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CNTNAP1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR