Stampa…

Anti-LRRC4B Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Fornitore: Bioss
BOSSBS-11090R-A350EA 858 EUR
BOSSBS-11090R-A350
Anti-LRRC4B Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorpi
LRRC4B/NGL3

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LRRC4B
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR