Stampa…

Anti-SPON2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fornitore: Bioss
BOSSBS-11064R-A555EA 858 EUR
BOSSBS-11064R-A555
Anti-SPON2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorpi
The Thrombospondin proteins, Thrombospondins 1-4 and Thrombospondin 5 (also designated COMP), compose a family of glycoproteins that are involved in cell-to-cell and cell-to-matrix signaling. These extracellular, cell-surface proteins form complexes of both homo- and heteromultimers. Spondin-2, or Mindin, is also designated DIL-1 for its differential expression in cancerous and non-cancerous lung cells. Full-length SPON2 cDNA encodes a 331 amino acid protein with a domain arrangement similar to zebrafish F-Spondin and Mindin-1/Mindin-2: an FS1 domain, an FS2 domain, a hydrophobic signal sequence in the N-terminus and a Thrombospondin type I repeat. Immunoblot analysis demonstrates expression of dimers and oligomers in a concentration-dependent manner under nonreducing conditions.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SPON2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR