Stampa…

Anti-HAPLN2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Fornitore: Bioss
BOSSBS-11051R-A488EA 858 EUR
BOSSBS-11051R-A488
Anti-HAPLN2 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorpi
HAPLN2 is a 340 amino acid protein encoded by the human gene HAPLN2. HAPLN2 belongs to the HAPLN family and contains one immunoglobulin (Ig)-like, V-type domain and two link domains. HAPLN2 mediates a firm binding of versican V2 to hyaluronic acid. HAPLN2 is believed to play a pivotal role in the formation of the hyaluronan-associated matrix in the central nervous system (CNS), which facilitates neuronal conduction and general structural stabilization. HAPLN2 may also be involved in the formation of extracellular matrices, contributing to perineuronal nets and facilitating the understanding of a functional role of these extracellular matrices. HAPLN2 is found in several nuclei throughout the midbrain and hindbrain in a perineuronal net pattern.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HAPLN2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR