Stampa…

Anti-PAX9 Rabbit Polyclonal Antibody (Cy5.5®)

Fornitore: Bioss
BOSSBS-1102R-CY5.5EA 858 EUR
BOSSBS-1102R-CY5.5
Anti-PAX9 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorpi
Transcription factor required for normal development of thymus, parathyroid glands, ultimobranchial bodies, teeth, skeletal elements of skull and larynx as well as distal limbs.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PAX9
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR