Stampa…

Anti-MYBPC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Fornitore: Bioss
BOSSBS-11034R-A647EA 858 EUR
BOSSBS-11034R-A647
Anti-MYBPC1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorpi
MYBPC1 is a 1,141 amino acid protein that contains three fibronectin type-III domains and seven Ig-like C2-type domains. Existing as a member of the immunoglobulin superfamily, MYBPC1 functions as a thick filament-associated protein that localizes to striated muscle bands in vertebrae and is thought to modify the activity of select ATPases. Additionally, MYBPC1 may play a role in the modulation of muscle contraction and in the overall structural integrity of the cell. The gene encoding MYBPC1 maps to human chromosome 12, which encodes over 1,100 genes and comprises approximately 4.5% of the human genome. Chromosome 12 is associated with a variety of diseases and afflictions, including hypochondrogenesis, achondrogenesis, Kniest dysplasia, Noonan syndrome and Trisomy 12p, which causes facial developmental defects and seizure disorders.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MYBPC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR