Stampa…

Anti-DNAH10 Rabbit Polyclonal Antibody (Cy3®)

Fornitore: Bioss
BOSSBS-11022R-CY3EA 858 EUR
BOSSBS-11022R-CY3
Anti-DNAH10 Rabbit Polyclonal Antibody (Cy3®)
Anticorpi
DNAH10

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: DNAH10
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR