Stampa…

Anti-HCRTR2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Fornitore: Bioss
BOSSBS-1095R-A647EA 858 EUR
BOSSBS-1095R-A647
Anti-HCRTR2 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorpi
The protein encoded by this gene is a G-protein coupled receptor involved in the regulation of feeding behavior. The encoded protein binds the hypothalamic neuropeptides orexin A and orexin B. A related gene (HCRTR1) encodes a G-protein coupled receptor that selectively binds orexin A. [provided by RefSeq, Jan 2009]

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HCRTR2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR