Stampa…

Anti-NOX4 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BOSSBS-1091R-CY7EA 866 EUR
BOSSBS-1091R-CY7
Anti-NOX4 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
Constitutive NADPH oxidase which generates superoxide intracellularly upon formation of a complex with CYBA/p22phox. Regulates signaling cascades probably through phosphatases inhibition. May function as an oxygen sensor regulating the KCNK3/TASK-1 potassium channel and HIF1A activity. May regulate insulin signaling cascade. May play a role in apoptosis, bone resorption and lipolysaccharide-mediated activation of NFKB.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOX4
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR