Stampa…

Anti-ANGPT4 Rabbit Polyclonal Antibody (Cy5.5®)

Fornitore: Bioss
BOSSBS-1087R-CY5.5EA 842 EUR
BOSSBS-1087R-CY5.5
Anti-ANGPT4 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorpi
Binds to TEK/TIE2, modulating ANGPT1 signaling. Can induce tyrosine phosphorylation of TEK/TIE2. Promotes endothelial cell survival, migration and angiogenesis.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ANGPT4
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR