Stampa…

Anti-KIF15 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BOSSBS-10693R-CY7EA 858 EUR
BOSSBS-10693R-CY7
Anti-KIF15 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
KIF15 belongs to the kinesin-like protein family and contains 1 kinesin-motor domain. KIF15 is the plus-end directed kinesin-like motor enzyme involved in mitotic spindle assembly.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KIF15
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR