Stampa…

Anti-NCAN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornitore: Bioss
BOSSBS-1055R-FITCEA 866 EUR
BOSSBS-1055R-FITC
Anti-NCAN Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
May modulate neuronal adhesion and neurite growth during development by binding to neural cell adhesion molecules (NG-CAM and N-CAM). Chondroitin sulfate proteoglycan; binds to hyaluronic acid.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NCAN
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR