Stampa…

Anti-GJA5 Rabbit Polyclonal Antibody (Cy5.5®)

Fornitore: Bioss
BOSSBS-1050R-CY5.5EA 866 EUR
BOSSBS-1050R-CY5.5
Anti-GJA5 Rabbit Polyclonal Antibody (Cy5.5®)
Anticorpi
One gap junction consists of a cluster of closely packed pairs of transmembrane channels, the connexons, through which materials of low MW diffuse from one cell to a neighboring cell.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Connexin-40
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Dog
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR