Stampa…

Anti-POMC Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BOSSBS-0443R-CY7EA 858 EUR
BOSSBS-0443R-CY7
Anti-POMC Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
ACTH stimulates the adrenal glands to release cortisol. MSH (melanocyte-stimulating hormone) increases the pigmentation of skin by increasing melanin production in melanocytes. Beta-endorphin and Met-enkephalin are endogenous opiates.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: POMC
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR