Stampa…

Anti-CHRM2 Rabbit Polyclonal Antibody (Cy5®)

Fornitore: Bioss
BOSSBS-0441R-CY5EA 866 EUR
BOSSBS-0441R-CY5
Anti-CHRM2 Rabbit Polyclonal Antibody (Cy5®)
Anticorpi
The muscarinic acetylcholine receptor mediates various cellular responses, including inhibition of adenylate cyclase, breakdown of phosphoinositides and modulation of potassium channels through the action of G proteins. Primary transducing effect is adenylate cyclase inhibition. Signaling promotes phospholipase C activity, leading to the release of inositol trisphosphate (IP3); this then triggers calcium ion release into the cytosol.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CHRM2
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR