Stampa…

Anti-Streptavidin Rabbit Polyclonal Antibody (Cy3®)

Fornitore: Bioss
BOSSBS-0437R-CY3EA 842 EUR
BOSSBS-0437R-CY3
Anti-Streptavidin Rabbit Polyclonal Antibody (Cy3®)
Anticorpi
Streptavidin is biotin-binding protein that was originally isolated from Streptomyces avidinii. In contrast to avidin, streptavidin has no carbohydrate and has a mildly acidic pI of 5. Streptavidin products use a recombinant form of streptavidin having a mass of 53,000 daltons and a near-neutral pI. Streptavidin is a tetrameric protein, with each subunit binding one molecule of biotin with affinity similar to that of avidin. Guanidinium chloride will dissociate avidin and streptavidin into subunits, but streptavidin is more resistant to dissociation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: Streptavidin
Clonality: Polyclonal
Clone:
Conjugation: Cy3®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR