Stampa…

Anti-TIMP4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)

Fornitore: Bioss
BOSSBS-0418R-A555EA 858 EUR
BOSSBS-0418R-A555
Anti-TIMP4 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Anticorpi
Complexes with metalloproteinases (such as collagenases) and irreversibly inactivates them by binding to their catalytic zinc cofactor. Known to act on MMP-1, MMP-2, MMP-3, MMP-7 and MMP-9.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: TIMP4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR