Stampa…

Anti-CRH Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Fornitore: Bioss
BOSSBS-0382R-A488EA 858 EUR
BOSSBS-0382R-A488
Anti-CRH Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Anticorpi
This hormone from hypothalamus regulates the release of corticotropin from pituitary gland.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CRH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR