Stampa…

Anti-NOS3 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BOSSBS-0356R-CY7EA 858 EUR
BOSSBS-0356R-CY7
Anti-NOS3 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
Digoxin is a beta blocker drug originally derived from the foxglove plant, Digitalis lanata. Digoxin is used primarily to improve the pumping ability of the heart in congestive heart failure (CHF), and treat problems such as high blood pressure. It is also used to help normalize some dysrhythmias (abnormal types of heartbeat).

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: NOS3
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Others
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR