Stampa…

Anti-BRI3BP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Fornitore: Bioss
BOSSBS-0402R-HRPEA 858 EUR
BOSSBS-0402R-HRP
Anti-BRI3BP Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Anticorpi
Involved in tumorigenesis and may function by stabilizing p53/TP53.

Recommended Dilutions: Western Blot: 1:100-1000; IHC-P: 1:100-500

Type: Primary
Antigen: BRI3BP
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR