Stampa…

Anti-BRI3BP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Fornitore: Bioss
BOSSBS-0402R-FITCEA 858 EUR
BOSSBS-0402R-FITC
Anti-BRI3BP Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Anticorpi
Involved in tumorigenesis and may function by stabilizing p53/TP53.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: BRI3BP
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR