Stampa…

Anti-HGV Polyprotein Rabbit Polyclonal Antibody (Cy5®)

Fornitore: Bioss
BOSSBS-0338R-CY5EA 858 EUR
BOSSBS-0338R-CY5
Anti-HGV Polyprotein Rabbit Polyclonal Antibody (Cy5®)
Anticorpi
Hepatitis G virus is from a group of viruses called GB viruses (GB virus A, GB virus B and GB virus C) and is also designated as GB virus C (GBV C). HGV is a positive stranded RNA virus, with a genome of 10 KB and appears, on the basis of genome length and organisation, to be a member of the Flaviviridae and distinct from the hepatitis C virus (HCV). HGV infection appears to be transmitted following receipt of blood from HGV RNA positive donors, however, these patients have only moderate liver enzyme elevations and do not develop chronic liver disease.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HGV Polyprotein
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR