Stampa…

Anti-FGF7 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BOSSBS-0734R-CY7EA 866 EUR
BOSSBS-0734R-CY7
Anti-FGF7 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: KGF
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR