Stampa…

Anti-SERPINF1 Rabbit Polyclonal Antibody (Cy7®)

Fornitore: Bioss
BOSSBS-0731R-CY7EA 842 EUR
BOSSBS-0731R-CY7
Anti-SERPINF1 Rabbit Polyclonal Antibody (Cy7®)
Anticorpi
Neurotrophic protein; induces extensive neuronal differentiation in retinoblastoma cells. Potent inhibitor of angiogenesis. As it does not undergo the S (stressed) to R (relaxed) conformational transition characteristic of active serpins, it exhibits no serine protease inhibitory activity.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: SERPINF1
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR