Stampa…

Anti-CXORF36 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Fornitore: Bioss
BOSSBS-0727R-A647EA 858 EUR
BOSSBS-0727R-A647
Anti-CXORF36 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Anticorpi
CXorf36

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CXORF36
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR