Stampa…

Anti-COL2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Fornitore: Bioss
BOSSBS-0709R-A350EA 866 EUR
BOSSBS-0709R-A350
Anti-COL2A1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Anticorpi
Type II collagen is specific for cartilaginous tissues. It is essential for the normal embryonic development of the skeleton, for linear growth and for the ability of cartilage to resist compressive forces.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: COL2A1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR