- Antibody type:Primary
- Antigen symbol:PBEF CT
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
icotinamide phosphoribosyltransferase (NAmPRTase) (Nampt) (Pre-B cell-enhancing factor) (Pre-B-cell colony-enhancing factor 1) (Fisfatin) Catalyzes the condensation of nicotinamide with5-phosphoribosyl-1-pyrophosphate to yield nicotinamide mononucleotide, an intermediate in the biosynthesis of NAD. It is the rate limiting component in the mammalian NAD biosynthesis pathway. Expressed in large amounts in bone marrow, liver tissue, and muscle. Also present in heart, placenta, lung, and kidney tissues. Subcellular Location is in Cytoplasm. Belongs to the NAPRTase family.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: PBEF CT
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat