Anti-TH Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-0016R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Tyrosine Hydroxylase
- Antigen symbol:TH
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:7054
- Antigen synonyms:TH-4|tyrosine hydroxylase|EC 1.14.16.2|TYH|Tyrosine 3-hydroxylase|Tyrosine 3-monooxygenase|TY3H|TH isoform 3|TH isoform a
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:115-160/528
- Purification:Purified by Protein A
- Pk:100 µl
Neuronal Marker Tyrosine hydroxylase is involved in the conversion of tyrosine to dopamine. As the rate-limiting enzyme in the synthesis of catecholamines, tyrosine hydroxylase has a key role in the physiology of adrenergic neurons. Tyrosine hydroxylase is regularly used as a marker for dopaminergic neurons, which is particularly relevant for research into Parkinson's disease.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: TH
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat