Skip to Header
Skip to Main Content
Skip to Footer
/
Italy
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Home
Buffers
Electrophoresis Buffers
Running Buffers
SDS-Page running gel buffers, 10X concentrate
SDS-Page running gel buffers, 10X concentrate
SDS-Page running gel buffers, 10X concentrate
ROCKMB-017
:
Rockland Immunochemicals
:
SDS-Page running gel buffers, 10X concentrate
ROCKMB-017
:
Rockland Immunochemicals
:
195,00 €
1 * 1 L
-
+
Pk:
1 L
Description:
SDS-PAGE 10X running gel buffer (0,25 M tris, 1,92 M glycine, 1,0% SDS pH 8,3)
Seq:
08